Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitrothiophene-2-carboxylic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
6317-37-9
C5H3NO4S
173.142
MFCD00159552
UNEPVPOHGXLUIR-UHFFFAOYSA-N
5-nitrothiophene-2-carboxylic acid,5-nitro-2-thiophenecarboxylic acid,5-nitro-2-thenoic acid,5-nitrothiophene-2-carboxylicacid,5-nitro-thiophene-2-carboxylic acid,2-thiophenecarboxylic acid, 5-nitro,2-carboxy-5-nitrothiophene,pubchem5568,acmc-1b47f,5-nitrothiophene-2-carboxylic
80591
C1=C(SC(=C1)[N+](=O)[O-])C(=O)O
6317-37-9
C5H3NO4S
173.142
MFCD00159552
UNEPVPOHGXLUIR-UHFFFAOYSA-N
5-nitrothiophene-2-carboxylic acid,5-nitro-2-thiophenecarboxylic acid,5-nitro-2-thenoic acid,5-nitrothiophene-2-carboxylicacid,5-nitro-thiophene-2-carboxylic acid,2-thiophenecarboxylic acid, 5-nitro,2-carboxy-5-nitrothiophene,pubchem5568,acmc-1b47f,5-nitrothiophene-2-carboxylic
80591
C1=C(SC(=C1)[N+](=O)[O-])C(=O)O
Melting Point 154°C to 159°C
Color Yellow
Quantity 100g
Beilstein 139665
Formula Weight 173.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Nitrothiophene-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning