Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nonanol, 97%, Acros Organics™

Click to view available options
Quantity:
5mL
25mL
623-93-8
C9H20O
144.26
FCBBRODPXVPZAH-UHFFFAOYSA-N
5-nonanol,dibutylcarbinol,dibutyl carbinol,5-nonyl alcohol,nonanol-5,acmc-1bc2t,di-n-butyl carbinol,5-nonanol 25g,5-nonanol gc,5-nonanol, puriss gc
12202
nonan-5-ol
CCCCC(CCCC)O
623-93-8
C9H20O
144.26
FCBBRODPXVPZAH-UHFFFAOYSA-N
5-nonanol,dibutylcarbinol,dibutyl carbinol,5-nonyl alcohol,nonanol-5,acmc-1bc2t,di-n-butyl carbinol,5-nonanol 25g,5-nonanol gc,5-nonanol, puriss gc
12202
nonan-5-ol
CCCCC(CCCC)O
Specific Gravity 0.82
Infrared Spectrum Authentic
Refractive Index 1.429
Boiling Point 194.0°C to 196.0°C
Quantity 5mL
Density 0.8200g/mL
Melting Point 5.0°C
Flash Point 77°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Solubility Information Solubility in water: insoluble.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material 5-Nonanol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 210-820-2