Learn More
5-Nonyloxytryptamine oxalate, Tocris Bioscience™
Selective 5-HT1B agonist
Manufacturer:  Tocris Bioscience 0901/5
 View more versions of this product
Catalog No. 09-015
Add to cart
Description
Description
5-HT1B selective agonist, several times more potent than sumatriptan and inactive as a 5-HT1A agonist (Ki at 5-HT1B = 1 nM, selectivity over 5-HT1A > 300-fold). Mimics polysialic acid activity, stimulates neuritogenesis, myelination and Schwann cell migration in vitro.
Specifications
Specifications
157798-13-5
C19H30N2O.C2H2O4
5mg
>99%
Observed MW: 392.49
Safety and Handling
Safety and Handling
Recommended Storage : Store at Room Temperature