Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Norbornene-2-carboxaldehyde, endo + exo, 95%

Click to view available options
Quantity:
25g
100g
5453-80-5
C8H10O
122.167
MFCD00167568
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
5-norbornene-2-carboxaldehyde,bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde,bicyclo 2.2.1 hept-5-ene-2-carbaldehyde,2-formyl-5-norbornene,5-formylbicyclohept-2-ene,5-norbornene-2-carbaldehyde,5-norbornane-2-carboxaldehyde,bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
95117
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
C1C2CC(C1C=C2)C=O
5453-80-5
C8H10O
122.167
MFCD00167568
AJIBZRIAUXVGQJ-UHFFFAOYSA-N
5-norbornene-2-carboxaldehyde,bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde,bicyclo 2.2.1 hept-5-ene-2-carbaldehyde,2-formyl-5-norbornene,5-formylbicyclohept-2-ene,5-norbornene-2-carbaldehyde,5-norbornane-2-carboxaldehyde,bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
95117
bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
C1C2CC(C1C=C2)C=O
Applications
5-Norbornene-2-carboxaldehyde was used in transition metal-catalyzed synthesis of norbornene-derived homopolymers having pendent t-Boc-protected quinizarin moieties for patterned fluorescence images. It acts as Diels-Alder adduct of acrolein and cyclopentadiene. It can be used as protected form of acrolein, from which the double bond may be liberated by cycloreversion, e.g. in a useful synthesis of 2-vinylimidazole.

Solubility
Not miscible in water.

Notes
Store away from air and oxidizing agents. Airsensitive. Store under dry inert gas.
Boiling Point 68°C to 70°C (12mmHg)
Flash Point 50°C (122°F)
Quantity 25g
Odor Characteristic
UN Number UN1989
Beilstein 1363813
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, endo + exo

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

TSCA : TSCA