Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornene-2-carboxylic acid, 98%, mixture of isomers, Acros Organics

Click to view available options
Quantity:
5g
100g
120-74-1
C8H10O2
138.16
FYGUSUBEMUKACF-UHFFFAOYSA-N
5-norbornene-2-carboxylic acid,bicyclo 2.2.1 hept-5-ene-2-carboxylic acid,norbornenecarboxylic acid,exo-5-norbornene-2-carboxylic acid,5-norbornene-2-carboxylic acid, exo,5-norbornene-2-carboxylic acid 8ci,bicyclo 2.2.1 hept-2-ene-5-carboxylic acid,bicyclo 2.2.1-5-heptene-2-carboxylic acid,norborn-5-ene-2-carboxylic acid
78949
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
C1C2CC(C1C=C2)C(=O)O
120-74-1
C8H10O2
138.16
FYGUSUBEMUKACF-UHFFFAOYSA-N
5-norbornene-2-carboxylic acid,bicyclo 2.2.1 hept-5-ene-2-carboxylic acid,norbornenecarboxylic acid,exo-5-norbornene-2-carboxylic acid,5-norbornene-2-carboxylic acid, exo,5-norbornene-2-carboxylic acid 8ci,bicyclo 2.2.1 hept-2-ene-5-carboxylic acid,bicyclo 2.2.1-5-heptene-2-carboxylic acid,norborn-5-ene-2-carboxylic acid
78949
bicyclo[2.2.1]hept-2-ene-5-carboxylic acid
C1C2CC(C1C=C2)C(=O)O
Specific Gravity 1.129
Density 1.1290g/mL
Boiling Point 136.0°C to 138.0°C (14.0mmHg)
Infrared Spectrum Authentic
Quantity 5g
Flash Point >110°C
Assay Percent Range 97.5% min. (total of isomers) (GC)
Formula Weight 138.16
Percent Purity 98%
Chemical Name or Material 5-Norbornene-2-carboxylic acid

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 204-422-8