Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 ft.-Nucleotidase/CD73 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1G2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271214JF646

Catalog No. N271214J646


Add to cart

Description

Description

5 ft.-Nucleotidase/CD73 Monoclonal antibody specifically detects 5 ft.-Nucleotidase/CD73 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

5′-Nucleotidase/CD73
OTI1G2
Immunohistochemistry
Affinity Purified, Conjugated
NT5E
Recombinant protein fragment corresponding to amino acids 119-232 of human 5′-Nucleotidase/CD73 (NP_002517) produced in E.coli.
Immunogen affinity purified
Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary
4907.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit