Learn More
5 ft.-Nucleotidase/CD73 Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 45M3B4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225235AF350
Catalog No. NP2223A30
Add to cart
Description
Description
5 ft.-Nucleotidase/CD73 Monoclonal antibody specifically detects 5 ft.-Nucleotidase/CD73 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
5′-Nucleotidase/CD73
45M3B4
Western Blot, Flow Cytometry
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
This product can detect Isoform 1 and Isoform of Human CD73.
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
NT5E
Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
0.1 mL
Primary
4907
Human, Mouse