Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 ft.-Nucleotidase/CD73 Mouse anti-Human, Mouse, Alexa Fluor™ 594, Clone: 45M3B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225235AF594

Catalog No. NP2223A94


Add to cart

Description

Description

5 ft.-Nucleotidase/CD73 Monoclonal antibody specifically detects 5 ft.-Nucleotidase/CD73 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

5′-Nucleotidase/CD73
45M3B4
Western Blot, Flow Cytometry
Conjugated, Purified
NT5E
Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Protein G purified
Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary
4907.0
Human, Mouse
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This product can detect Isoform 1 and Isoform of Human CD73.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit