Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5′-Nucleotidase/CD73 Rabbit anti-Human, Clone: 102, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | 5′-Nucleotidase/CD73 |
---|---|
Clone | 102 |
Dilution | ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
5′-Nucleotidase/CD73 | |
ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
Recombinant Human 5′-Nucleotidase/CD73 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
102 | |
ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Myeloid derived Suppressor Cell | |
0.2 um filtered solution in PBS | |
4907 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title