Learn More
5'-Nucleotidase/CD73 Rat anti-Mouse, Alexa Fluor 405, Clone: 496406, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB4488V100UG
Catalog No. FAB4488V100
Add to cart
This item is not returnable. View return policy
Description
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
5'-Nucleotidase/CD73
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q61503
NT5E
Chinese hamster ovary cell line CHO-derived recombinant mouse 5'-Nucleotidase/CD73 Trp29-Lys549 Accession # Q61503
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse 5'-Nucleotidase/CD73 in direct ELISAs.
Flow Cytometry
496406
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
5' nucleotidase (CD73), 5'-NT, 5-NT, 5'Nucleotidase, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NT5E, NTE, Purine 5-Prime-Nucleotidase
Rat
IgG2a
RUO
Primary
4907
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)