Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 Percent Palladium on Activated Carbon Powder, Reduced, Dry, 27 microns, BASF Catalyst, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
7440-05-3
Pd
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
[Pd]
7440-05-3
Pd
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
[Pd]
Additional Information Support: Activated Carbon powder
Palladium (Pd) 0.05
Particle Size 27 microns
Quantity 5 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.