Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-1-pentanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
10521-91-2
C11H16O
164.25
MFCD00002979
DPZMVZIQRMVBBW-UHFFFAOYSA-N
5-phenyl-1-pentanol,benzenepentanol,5-phenylpentanol,phenylamyl alcohol,phenylpentanol,1-pentanol, 5-phenyl,pentanol, 5-phenyl,phenylpentanol van,unii-63ax6o7zxd,5-phenyl-pentanol
61523
5-phenylpentan-1-ol
C1=CC=C(C=C1)CCCCCO
10521-91-2
C11H16O
164.25
MFCD00002979
DPZMVZIQRMVBBW-UHFFFAOYSA-N
5-phenyl-1-pentanol,benzenepentanol,5-phenylpentanol,phenylamyl alcohol,phenylpentanol,1-pentanol, 5-phenyl,pentanol, 5-phenyl,phenylpentanol van,unii-63ax6o7zxd,5-phenyl-pentanol
61523
5-phenylpentan-1-ol
C1=CC=C(C=C1)CCCCCO
Flash Point >110°C
Quantity 5g
Color Colorless to Yellow
Boiling Point 155.0°C
Refractive Index 1.5162
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5(CH2)5OH
Beilstein 06, IV, 3373
Formula Weight 164.25
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 5-Phenyl-1-pentanol, 98%
Show More Show Less

EINECSNumber : 234-064-8