Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenyl-o-anisidine, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
39811-17-1
C13H13NO
199.253
MFCD00007790
DTYBRSLINXBXMP-UHFFFAOYSA-N
5-phenyl-o-anisidine,3-amino-4-methoxybiphenyl,4-methoxy-1,1'-biphenyl-3-amine,2-amino-4-phenylanisole,4-methoxybiphenyl-3-amine,1,1'-biphenyl-3-amine, 4-methoxy,2-methoxy-5-phenylphenylamine,acmc-209j7y,2-methoxy-5-phenyl-aniline,3-amino-4-methoxy-biphenyl
123489
2-methoxy-5-phenylaniline
COC1=C(C=C(C=C1)C2=CC=CC=C2)N
39811-17-1
C13H13NO
199.253
MFCD00007790
DTYBRSLINXBXMP-UHFFFAOYSA-N
5-phenyl-o-anisidine,3-amino-4-methoxybiphenyl,4-methoxy-1,1'-biphenyl-3-amine,2-amino-4-phenylanisole,4-methoxybiphenyl-3-amine,1,1'-biphenyl-3-amine, 4-methoxy,2-methoxy-5-phenylphenylamine,acmc-209j7y,2-methoxy-5-phenyl-aniline,3-amino-4-methoxy-biphenyl
123489
2-methoxy-5-phenylaniline
COC1=C(C=C(C=C1)C2=CC=CC=C2)N
Infrared Spectrum Authentic
Melting Point 81°C to 84°C
Packaging Glass bottle
Color Brown
Quantity 5g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H5C6H3(OCH3)NH2
Beilstein 13, III, 1946
Formula Weight 199.25
Physical Form Coarse Powder
Percent Purity ≥98.0%
Chemical Name or Material 5-Phenyl-o-anisidine, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-639-7