Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Phenylisoxazole-3-carboxylic acid, ≥99%

Click to view available options
Quantity:
1g
5g
14441-90-8
C10H7NO3
189.17
MFCD00464221
XJYOBHXWBRKOQO-UHFFFAOYSA-N
5-phenylisoxazole-3-carboxylic acid,5-phenyl-isoxazole-3-carboxylic acid,5-phenyl-3-isoxazolecarboxylic acid,3-isoxazolecarboxylic acid, 5-phenyl,3-isoxazolecarboxylicacid, 5-phenyl,5-phenylisoxazole-3-carboxylicacid,5-phenylisoxazol-3-carboxylic acid,5-phenylisoxazole-3-carboxylic acid
151916
5-phenyl-1,2-oxazole-3-carboxylic acid
C1=CC=C(C=C1)C2=CC(=NO2)C(=O)O
14441-90-8
C10H7NO3
189.17
MFCD00464221
XJYOBHXWBRKOQO-UHFFFAOYSA-N
5-phenylisoxazole-3-carboxylic acid,5-phenyl-isoxazole-3-carboxylic acid,5-phenyl-3-isoxazolecarboxylic acid,3-isoxazolecarboxylic acid, 5-phenyl,3-isoxazolecarboxylicacid, 5-phenyl,5-phenylisoxazole-3-carboxylicacid,5-phenylisoxazol-3-carboxylic acid,5-phenylisoxazole-3-carboxylic acid
151916
5-phenyl-1,2-oxazole-3-carboxylic acid
C1=CC=C(C=C1)C2=CC(=NO2)C(=O)O
Quantity 5g
Melting Point 160°C to 164°C
Color White
Beilstein 146162
Formula Weight 189.17
Physical Form Crystalline
Percent Purity ≥99%
Chemical Name or Material 5-Phenylisoxazole-3-carboxylic acid

GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger