Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Propionyl-2,2'-bithienyl, 95+%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
32358-91-1
C11H10OS2
222.32
MFCD04039750
MRWPBEHIJTUNGC-UHFFFAOYSA-N
5-propionyl-2,2'-bithienyl,1-2,2'-bithiophen-5-yl propan-1-one,5-propionyl-2,2'-bithiophene,1-propanone,1-2,2'-bithiophen-5-yl,1-5-2-thienyl-2-thienyl propan-1-one,1-5-thiophen-2-ylthiophen-2-yl propan-1-one,1-5-thiophen-2-yl thiophen-2-yl propan-1-one
2734305
1-(5-thiophen-2-ylthiophen-2-yl)propan-1-one
CCC(=O)C1=CC=C(S1)C2=CC=CS2
32358-91-1
C11H10OS2
222.32
MFCD04039750
MRWPBEHIJTUNGC-UHFFFAOYSA-N
5-propionyl-2,2'-bithienyl,1-2,2'-bithiophen-5-yl propan-1-one,5-propionyl-2,2'-bithiophene,1-propanone,1-2,2'-bithiophen-5-yl,1-5-2-thienyl-2-thienyl propan-1-one,1-5-thiophen-2-ylthiophen-2-yl propan-1-one,1-5-thiophen-2-yl thiophen-2-yl propan-1-one
2734305
1-(5-thiophen-2-ylthiophen-2-yl)propan-1-one
CCC(=O)C1=CC=C(S1)C2=CC=CS2
Packaging Glass Bottle
Melting Point 87°C to 91°C
Color Green
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Formula Weight 222.33
Physical Form Crystalline Powder
Percent Purity ≥95.0%
Chemical Name or Material 5-Propionyl-2, 2'-bithienyl
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.