Learn More
Cayman ChemicalSupplier Diversity Partner 15(R)-Lipoxin A4, 171030-11-8, 50 µg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 9041550
An aspirin-triggered lipoxin that inhibits LTB4-induced chemotaxis, adherence, and transmigration of the neutrophils with twice the potency of LXA4 demonstrating activity in the nM range
Catalog No. NC0136505