Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Sulfosalicylic Acid Dihydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
5965-83-3,97-05-2
C7H6O62H2O
254.22
MFCD00149540
BHDKTFQBRFWJKR-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd
2723734
2-hydroxy-5-sulfobenzoic acid;dihydrate
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O.O.O
5965-83-3,97-05-2
C7H6O62H2O
254.22
MFCD00149540
BHDKTFQBRFWJKR-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid dihydrate,5-sulfosalicylic acid dihydrate,sulfosalicylic acid dihydrate,benzoic acid, 2-hydroxy-5-sulfo-, dihydrate,unii-09ngq462s6,2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate,5-sulfosalicylsyre,5-sulfosalicylsaeure,salicylic acid, 5-sulfo-, dihydrate,acmc-1aymd
2723734
2-hydroxy-5-sulfobenzoic acid;dihydrate
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O.O.O
Indicator
Quantity 2.5kg
Packaging Plastic bottle
Melting Point 106.0°C to 110.0°C
Color White
Infrared Spectrum Authentic
Water 14 to 16% (K.F.) (typically)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 11, 411
Fieser 01,1118
Merck Index 15, 9094
Solubility Information Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 254.22
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Sulfosalicylic acid dihydrate
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage. Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger