Learn More
Aat BioquestSupplier Diversity Partner 5-TAMRA [5-Carboxytetramethylrhodamine] *CAS 91809-66-4* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 365
5-TAMRA is the purified single isomer of 5(6)-TAMRA. It is widely used to label peptides and proteins.
Catalog No. 50-195-2854