Learn More
Aat BioquestSupplier Diversity Partner 5-TAMRA alkyne DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 961
5-TAMRA alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
Catalog No. 50-195-2907