Learn More
Aat BioquestSupplier Diversity Partner 5-TAMRA azide DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 960
5-TAMRA azide is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
Catalog No. 50-195-2905