Learn More
Aat BioquestSupplier Diversity Partner 5-TAMRA, SE [5-Carboxytetramethylrhodamine, succinimidyl ester] *CAS#: 150810-68-7* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 374
The single TAMRA isomers are increasingly preferred for labeling peptides and nucleotides because they give better resolution in HPLC purification that is often required in the conjugation processes.
Catalog No. 50-195-2862