Alfa Aesar™ 5-tert-Butyl-2-iodo-1,3-dimethylbenzene, 97%

Manufacturer: Alfa Aesar™ B2519514

Quantity 25g

 View more versions of this product

Catalog No. AAB2519514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5122-20-3
    Molecular Formula C12H17I
    Formula Weight 288.17
    MDL Number MFCD00052173
    Synonym 5-tert-Butyl-2-iodo-m-xylene
    Chemical Name or Material 5-tert-Butyl-2-iodo-1,3-dimethylbenzene

    Specifications

    Formula Weight 288.17
    Synonym 5-tert-Butyl-2-iodo-m-xylene
    Odor Odorless
    Quantity 25g
    Molecular Formula C12H17I
    MDL Number MFCD00052173
    CAS 5122-20-3
    Melting Point 54° to 57°C
    Chemical Name or Material 5-tert-Butyl-2-iodo-1,3-dimethylbenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit