Learn More
Medchemexpress LLC 5-(tetradecyloxy)-2-furoic acid | 54857-86-2 | MFCD01726059 | 99.9% | 324.45 g·mol⁻¹ | C19H32O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10106825MG
Encompass_Preferred
TOFA (5-(tetradecyloxy)-2-furoic acid) is an allosteric inhibitor of acetyl-CoA carboxylase used as a research reagent to block fatty acid synthesis in biochemical and cell biology studies. It is cell-permeable and commonly applied in vitro to investigate lipid metabolism and related signaling pathways. For research use only; not for human or veterinary applications.
  • Allosteric inhibitor of acetyl-CoA carboxylase.
  • Reduces cellular fatty acid synthesis.
  • Cell-permeable small molecule suitable for in vitro assays.
  • Supplied in laboratory-ready quantities for experimental use.
  • High purity for reproducible biochemical results.
Catalog No. 50-002-12589