Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Tri-n-butylstannyl)thiazole, 96%

Manufacturer:  Alfa Aesar™ H5145503

Quantity 1g

 View more versions of this product

Catalog No. AAH5145503

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

157025-33-7
374.17
UN2788
5-(Tri-n-butylstannyl)thiazole
C15H29NSSn
MFCD01319074
5-Thiazolyltri-n-butylstannane; (Tri-n-butyl)-5-thiazolyltin

Specifications

1g
C15H29NSSn
MFCD01319074
5-Thiazolyltri-n-butylstannane; (Tri-n-butyl)-5-thiazolyltin
96%
157025-33-7
374.17
UN2788
5-(Tri-n-butylstannyl)thiazole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P273-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit