Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Triethylsilyl-4-pentyn-1-ol +97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
174064-02-9
C11H22OSi
198.381
MFCD00671375
RDNBUIIPNLWRPB-UHFFFAOYSA-N
5-triethylsilyl-4-pentyn-1-ol,5-triethylsilyl pent-4-yn-1-ol,4-pentyn-1-ol,5-triethylsilyl,5-triethylsilyl-4-pentin-1-ol
2760695
5-triethylsilylpent-4-yn-1-ol
CC[Si](CC)(CC)C#CCCCO
174064-02-9
C11H22OSi
198.381
MFCD00671375
RDNBUIIPNLWRPB-UHFFFAOYSA-N
5-triethylsilyl-4-pentyn-1-ol,5-triethylsilyl pent-4-yn-1-ol,4-pentyn-1-ol,5-triethylsilyl,5-triethylsilyl-4-pentin-1-ol
2760695
5-triethylsilylpent-4-yn-1-ol
CC[Si](CC)(CC)C#CCCCO
Packaging Glass bottle
Density 0.8600g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 5g
Refractive Index 1.4680 to 1.4700 (20°C, 589nm)
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula (CH3CH2)3SiCidentical toC(CH2)3OH
Formula Weight 198.38
Physical Form Liquid
Percent Purity ≥97.0%
Chemical Name or Material 5-Triethylsilyl-4-pentyn-1-ol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.