Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Trifluoromethoxy)salicylaldehyde, 98+%

Click to view available options
Quantity:
250mg
1g
93249-62-8
C8H5F3O3
206.12
MFCD00075249
WQUZBERVMUEJTD-UHFFFAOYSA-N
2-hydroxy-5-trifluoromethoxy benzaldehyde,5-trifluoromethoxy salicylaldehyde,benzaldehyde, 2-hydroxy-5-trifluoromethoxy,acmc-20antc,5-trifluoromethoxysalicylaldehyde,5-trifluoromethoxy-salicylaldehyde,2-hydroxy-5-trifluoromethoxy-benzaldehyde,20hydroxy-5-trifluoromethoxy benzaldehyde,2-oxidanyl-5-trifluoromethyloxy benzaldehyde,benzaldehyde,2-hydroxy-5-trifluoromethoxy
1268058
2-hydroxy-5-(trifluoromethoxy)benzaldehyde
C1=CC(=C(C=C1OC(F)(F)F)C=O)O
93249-62-8
C8H5F3O3
206.12
MFCD00075249
WQUZBERVMUEJTD-UHFFFAOYSA-N
2-hydroxy-5-trifluoromethoxy benzaldehyde,5-trifluoromethoxy salicylaldehyde,benzaldehyde, 2-hydroxy-5-trifluoromethoxy,acmc-20antc,5-trifluoromethoxysalicylaldehyde,5-trifluoromethoxy-salicylaldehyde,2-hydroxy-5-trifluoromethoxy-benzaldehyde,20hydroxy-5-trifluoromethoxy benzaldehyde,2-oxidanyl-5-trifluoromethyloxy benzaldehyde,benzaldehyde,2-hydroxy-5-trifluoromethoxy
1268058
2-hydroxy-5-(trifluoromethoxy)benzaldehyde
C1=CC(=C(C=C1OC(F)(F)F)C=O)O
Sensitivity Air Sensitive
Boiling Point 82°C (60mmHg)
Flash Point >110°C (230°F)
Quantity 250mg
Melting Point 31°C to 33°C
Beilstein 7023825
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 5-(Trifluoromethoxy)salicylaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen