Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog Number p-4090102
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
129946-88-9
C14H8F6O3S2
402.33
MFCD00236132
QXXHXTRTGZBOGD-UHFFFAOYSA-M
5-trifluoromethyl-5h-dibenzo b,d thiophen-5-ium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, 5-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium triflate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulphonate, trifluoromethanesulfonate; 5-trifluoromethyl dibenzothiophen-5-ium, pubchem16425, acmc-20a6nt, c13h8f3s.chf3o3s, s-trifluoromethyl dibenzothiophenium trifluoromet
2777507
trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12
129946-88-9
C14H8F6O3S2
402.33
MFCD00236132
QXXHXTRTGZBOGD-UHFFFAOYSA-M
5-trifluoromethyl-5h-dibenzo b,d thiophen-5-ium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, 5-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium triflate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulphonate, trifluoromethanesulfonate; 5-trifluoromethyl dibenzothiophen-5-ium, pubchem16425, acmc-20a6nt, c13h8f3s.chf3o3s, s-trifluoromethyl dibenzothiophenium trifluoromet
2777507
trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 150°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 402.33
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit