Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%

Click to view available options
Quantity:
1g
5g
25g
80194-69-0
C7H4F3NO2
191.109
MFCD04113632
NJHGVAYLDHROPT-UHFFFAOYSA-N
5-trifluoromethyl pyridine-2-carboxylic acid,5-trifluoromethyl picolinic acid,5-trifluoromethyl-2-pyridinecarboxylic acid,5-trifluoromethyl-2-pyridinecarboxylic acid,5-trifluoromethyl-pyridine-2-carboxylic acid,5-trifluoromethyl-pyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 5-trifluoromethyl,5-trifluoromethyl pyridine-2-carboxylicacid,pubchem15164,acmc-209pj2
10910349
5-(trifluoromethyl)pyridine-2-carboxylic acid
C1=CC(=NC=C1C(F)(F)F)C(=O)O
80194-69-0
C7H4F3NO2
191.109
MFCD04113632
NJHGVAYLDHROPT-UHFFFAOYSA-N
5-trifluoromethyl pyridine-2-carboxylic acid,5-trifluoromethyl picolinic acid,5-trifluoromethyl-2-pyridinecarboxylic acid,5-trifluoromethyl-2-pyridinecarboxylic acid,5-trifluoromethyl-pyridine-2-carboxylic acid,5-trifluoromethyl-pyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 5-trifluoromethyl,5-trifluoromethyl pyridine-2-carboxylicacid,pubchem15164,acmc-209pj2
10910349
5-(trifluoromethyl)pyridine-2-carboxylic acid
C1=CC(=NC=C1C(F)(F)F)C(=O)O
Applications
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Melting Point 133°C to 137°C
Color Brown
Quantity 25g
Formula Weight 191.11
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 5-(Trifluoromethyl)pyridine-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning