Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(5-Trifluoromethylpyridin-3-yl)boronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
947533-51-9
C6H5BF3NO2
190.92
MFCD09952041
SFBQNNGMEKUJAN-UHFFFAOYSA-N
5-trifluoromethyl-pyridine-3-boronic acid,5-trifluoromethyl pyridin-3-yl boronic acid,3-trifluoromethylpyridine-5-boronic acid,3-trifluoromethyl-5-pyridyl boronic acid,5-trifluoromethyl pyridine-3-boronic acid,5-trifluoromethyl pyridin-3-ylboronic acid,5-trifluoromethylpyridine-3-boronic acid,5-trifluoromethyl-3-pyridyl boronic acid,3-borono-5-trifluoromethyl pyridine
45158900
[5-(trifluoromethyl)pyridin-3-yl]boronic acid
B(C1=CC(=CN=C1)C(F)(F)F)(O)O
947533-51-9
C6H5BF3NO2
190.92
MFCD09952041
SFBQNNGMEKUJAN-UHFFFAOYSA-N
5-trifluoromethyl-pyridine-3-boronic acid,5-trifluoromethyl pyridin-3-yl boronic acid,3-trifluoromethylpyridine-5-boronic acid,3-trifluoromethyl-5-pyridyl boronic acid,5-trifluoromethyl pyridine-3-boronic acid,5-trifluoromethyl pyridin-3-ylboronic acid,5-trifluoromethylpyridine-3-boronic acid,5-trifluoromethyl-3-pyridyl boronic acid,3-borono-5-trifluoromethyl pyridine
45158900
[5-(trifluoromethyl)pyridin-3-yl]boronic acid
B(C1=CC(=CN=C1)C(F)(F)F)(O)O
Quantity 250mg
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 260.0°C to 262.6°C
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 190.92
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material (5-Trifluoromethylpyridin-3-yl)boronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning