Learn More
Cayman Chemical 5 Z 8 Z 11 ZEicosatrienoic Ac
SDP
Supplier:  Cayman Chemical 10742500 UG
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z),11(Z)-eicosatrienoic acid by GC- or LC-MS
Catalog No. 50-288-9236