Learn More
Exel International 50 to 60cc Syringes
Supplier: Exel International 26300
Description
Latex free;Sterile;Non-toxic;Pyrogen free;Positive plunger stop to prevent spillage when drawing;Smooth plunger motion;Polypropylene construction.
Specifications
Pyrogen Free | |
Syringe | |
Luer Lock |
Yes | |
50 to 60 mL | |
Sterile |