Learn More
TeknovaSupplier Diversity Partner 50 MM HEPES ASSAY DILUENT DFS Item
Manufacturer:  TeknovaSupplier Diversity Partner 2A3030
50mM HEPES Assay Diluent. pH 7.5, 1 Liter Bottle, Sterile.
Catalog No. NC0161929