Learn More
Agilent BioTek 50 TS Microplate Washer, IVD PROMO
Offers excellent microplate washing performance. For In Vitro Diagnostic Use.
Specifications
Certifications/Compliance | CE and TUV marked. RoHS Compliant. |
---|---|
Connections | USB (1) |
Bottles | Optional (2 to 4 L) |
Plate Type | 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) |
Soak Time | Programmable in minutes and seconds, up to 30 min. |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
50-TS1-2SIW25
|
Agilent BioTek
50TS12SI3044 |
50TS12 | N/A | N/A | N/A | ||||
50-TS1-6SIW25
|
Agilent BioTek
50TS16SI3044 |
50TS16 | N/A | N/A | N/A | ||||
50-TS8-FSIW25
|
Agilent BioTek
50TS8FSI3044 |
50TS8F | N/A | N/A | N/A | ||||
50-TS8-MSIW25
|
Agilent BioTek
50TS8MSI3044 |
50TS8M | N/A | N/A | N/A | ||||
50-TS8-SIW25
|
Agilent BioTek
50TS8SI3044 |
50TS8 | N/A | N/A | N/A | ||||
50-TS8-VSIW25
|
Agilent BioTek
50TS8VSI3044 |
50TS8V | N/A | N/A | N/A | ||||
5T-S12-VSIW25
|
Agilent BioTek
50TS12VSI3044 |
50TS12V | N/A | N/A | N/A | ||||
5T-S16-VSIW25
|
Agilent BioTek
50TS16VSI3044 |
50TS16V | N/A | N/A | N/A | ||||
5T-S8M-FSIW25
|
Agilent BioTek
50TS8MFSI3044 |
50TS8MF | N/A | N/A | N/A | ||||
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Specifications
CE and TUV marked. RoHS Compliant. | |
Optional (2 to 4 L) | |
Programmable in minutes and seconds, up to 30 min. | |
9.8 kg | |
25 to 3000 μL/well | |
150 to 1000 μL/well/sec. (Manifold Dependent) | |
4.3 in. Color LCD Touch Screen Display |
USB (1) | |
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) | |
Microplate Washer | |
1 to 10 | |
4.3 in. LCD Color Touch Screen | |
One Positive Displacement Syringe Drive | |
22 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.