Thermo Scientific™ 5000 Series 0.31µm to 0.93µm Polymer Particle Suspensions Available on GSA/VA Contract for Federal Government customers only.

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions

Manufacturer: Thermo Scientific™ 5045C

 View more versions of this product

Catalog No. 09-980-298

  Check Availability
Add to Cart

    Specifications

    Diameter (Metric) 0.45μm
    Type Specialty Particles
    For Use With (Application) Aerosol Particle Generation
    Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
    Description Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration.
    Diameter (Metric) Particle 0.45µm
    Includes General product handling insert sheet
    Packaging 1000mL
    Particle Density 1.05g/cm3
    Quantity 1000mL
    Refractive Index 1.59 at 589nm (25°C)
    Stability Product stable for a minimum of 24 months
    Storage Requirements Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
    Concentration 10%
    Density 1.05g/cm3

    Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions, of special interest for research in light scattering, microporous filter checking and aerosol particle generation.

    Features:

    • Density of 1.05g/cm3 and a refractive index of 1.59 at 589nm
    • Diameters measured by optical microscopy, photon correlation spectroscopy or light scattering
    • Packaged as aqueous suspensions at 10% solids by weight
    • Range up to 3.1 microns in size
    • For larger particles, refer to our 7000 Series Copolymer Particle Suspensions

    Ordering Information:

    • 15mL, 100mL, and 1000mL bottle sizes available
    • 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C).
    • Please allow 10 working days when ordering these fill volumes.

    Includes:

    Material Safety Data Sheet (MSDS) and general product handling insert sheet

    Recommended for:

    High concentration polymer particles < 3µm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit