Learn More
Athens Research And Technology IncSupplier Diversity Partner 500UG APOA II HUMAN PLASMA DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 1616120102500UG
Apolipoprotein AII, Human Plasma, HDL: 500 UG; Frozen in 10 mM NH4HCO3, pH 7.4; MW 17,400; Purity: > =95% by SDS-PAGE; This native protein purified from human plasma in the Athens Research & Tech lab. Molecular Biology Reagent, ApoLipoprotein
Catalog No. 50-489-630