Learn More
Leap PAL PartsSupplier Diversity Partner L-MARK Brand 500l Syringe  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner LMK.2620903
L-MARK Brand 500l, L-MARK Brand Syringe, FN, GT, 26gs, NDL Length 56mm, Pt. Style AS, Plunger Tip PTFE, Single Made in Germany.
Catalog No. 50-635-365