Learn More
ACROBiosystems Recombinant Protein;Human B7-H4 Protein;HEK293;100UG;B74,B7-H4,VTCN1,B7H4,B7S1,B7h.5,VTCN-1,B7-S1
Supplier:  ACROBiosystems B74H5222100UG
Encompass_Preferred
Recombinant Protein;Human B7-H4 Protein;100UG;Human B7-H4, His Tag (B74-H5222) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Ala 258 (Accession # NP_078902).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.1 kDa. The protein migrates as 43-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-H4, His Tag (Cat. No. B74-H5222) at 2 μg/mL can bind anti-B7-H4 mAb (Human IgG1) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-101-7237