Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human CD38 Protein;HEK293;50UG;CD8,CD38,T10,cADPr hydrolase 1
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CD8H522450UG
Encompass_Preferred
Recombinant Protein;Human CD38 Protein;50UG;Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # NP_001766).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD38, His Tag (Cat. No. CD8-H5224) at 5μg/mL (100 μL/well) can bind Anti-CD38 MAb, Human IgG1 with a linear range of 0.3-5.2 ng/mL (QC tested).
Catalog No. 50-101-7432