Learn More
ACROBiosystems Recombinant Protein;Human CD3 epsilon Protein;HEK293;100UG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Supplier: ACROBiosystems CDEH5223100UG
Recombinant Protein;Human CD3 epsilon Protein;100UG;Human CD3E, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.2 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 μg/mL (100 μL/well) can bind Human CD3E, His Tag (Cat. No. CDE-H5223) with a linear range of 0.4-6.25 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.