Learn More
ACROBiosystems Recombinant Protein;Human LSECtin / CLEC4G Protein, Fc Tag;HEK293;50UG;CLG,CLEC4G,LSECtin
Supplier:  ACROBiosystems CLGH525050UG
Encompass_Preferred
Recombinant Protein;Human LSECtin / CLEC4G Protein, Fc Tag;50UG;Human LSECtin, Fc Tag (CLG-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 55 - Cys 293 (Accession # Q6UXB4).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 52.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-7540