Learn More
ACROBiosystems Recombinant Protein;Human COMP / TSP5 Protein;HEK293;100UG;COP,COMP,TSP5,EDM1,EPD1,MED,PSACH,THBS5,TSP-5,Thrombospondin-5
Manufacturer:  ACROBiosystems COPH5220100UG
Encompass_Preferred
Recombinant Protein;Human COMP / TSP5 Protein;100UG;Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession # AAI25093).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 82 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-7553