Learn More
ACROBiosystems Recombinant Protein;Human CTLA-4 / CD152 Protein, Fc Tag;HEK293;200UG;CT4,CTLA4,CD152,CELIAC3,CTLA-4,GRD4,GSE,ICOS,IDDM12
Supplier:  ACROBiosystems CT4H5255200UG
Encompass_Preferred
Recombinant Protein;Human CTLA-4 / CD152 Protein, Fc Tag;200UG;Human CTLA-4, Fc Tag (CT4-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession # NP_005205.2).;This protein carries a human IgG1 Fc tag at the C-terminus, and has a calculated MW of 40 kDa. The predicted N-terminus is Ala 37. As a result of glycosylation, the reducing (R) protein migrates as 50-55 kDa, and the non-reducing (NR) protein migrates as 100-110 kDa in SDS-PAGE.;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 10 μg/mL (100 μL/well) can bind Human B7-1, His Tag (Cat. No. B71-H5228) with a linear range of 0.156-1.25 μg/mL (Routinely tested).;Immobilized Human B7-2, His Tag (Cat. No. CD6-H5223) at 10 μg/mL (100 µl/well), can bind Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) with a linear range of 0.04-0.3 μg/mL (QC tested).
Catalog No. 50-101-7596