Learn More
ACROBiosystems Recombinant Protein;Human E-Cadherin / Cadherin-1 Protein, Fc Tag;HEK293;100UG;ECD,Cadherin-1,CDH1,CD324,E-cadherin,Arc-1,ECAD,LCAM,UVO,E-CAD,Cadherin 1,CDH-1
Supplier:  ACROBiosystems ECDH5250100UG
Encompass_Preferred
Recombinant Protein;Human E-Cadherin / Cadherin-1 Protein, Fc Tag;100UG;Human E-Cadherin, Fc Tag (ECD-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Pro 621 (Accession # AAI41839).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.4 kDa. As a result of different glycosylation, the protein migrates as 110-125 kDa Cadherin, a 48kDa Propeptide under reducing (R) condition, and 200-250 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-101-7679
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.