Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human ErbB3 / Her3 Protein, His Tag;HEK293;100UG;ER3,ERBB3,ErbB-3,HER3,HER-3,LCCS2,MDA-BF-1,MGC88033,c-erbB-3,c-erbB3,erbB3-S,p180-ErbB3,p45-sErbB3,p85-sErbB3
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ER3H5223100UG
Encompass_Preferred
Recombinant Protein;Human ErbB3 / Her3 Protein, His Tag;100UG;Human ErbB3, His Tag (ER3-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Thr 643 (Accession # P21860-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-7739