Learn More
ACROBiosystems Recombinant Protein;Mouse FcRn/FCGRT & B2M Heterodimer Protein;HEK293;50UG;FCM,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Manufacturer:  ACROBiosystems FCMM52W250UG
Encompass_Preferred
Recombinant Protein;Mouse FcRn/FCGRT & B2M Heterodimer Protein;50UG;Mouse FcRn, His Tag, Strep Tag (FCM-M52W2) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Ser 297 (FCGRT) & Ile 21 - Met 119 (B2M) (Accession # Q61559 (FCGRT) & P01887 (B2M)).;Mouse FcRn / FCGRT & B2M Heterodimer Protein, produced by co-expression of FCGRT and B2M, has a calculated MW of 33 kDa (FCGRT) and 14.7 kDa (B2M). Subunit FCGRT is fused with his-tag at the C-terminus and subunit Beta-2 microglobulin (B2M) is fused with Twin-Strep tag at the C-terminus. The predicted N-terminus is Ser 22(FCGRT) & Ile 21(B2M). The reducing (R) protein migrates as 45-50 kDa (FCGRT) and 18 kDa (B2M) respectively due to glycosylation.
Catalog No. 50-101-7798