Learn More
ACROBiosystems Recombinant Protein;Human Frizzled-5 / FZD5 Protein, Fc Tag;HEK293;100UG;FZ5,FZD5,Frizzled-5,FZD-5,Frizzled 5,C2orf31,Fz-5,hFz5,FzE5,Frizzled 5
Manufacturer:  ACROBiosystems FZ5H5259100UG
Encompass_Preferred
Recombinant Protein;Human Frizzled-5 / FZD5 Protein, Fc Tag;100UG;Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-7847