Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade B/C (CN54)] GP120;HEK293;100UG;GP4,GP120,HIV,GP160,HIV-1,GP41,GP120-CN54,CN54,CN54-GP120  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner GP4V15227100UG
Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade B/C (CN54)] GP120;100UG;Recombinant HIV-1 GP120 Protein (HIV-1 /Clade B/C (CN54)) (Recombinant HIV-1 GP120 (CN54)) derived from the env. gene of HIV-1 strain CN54 gp160 (GenBank Gene Ac. # AX149771, Thr 27 - Arg 498) and glycosylated with N-linked sugars and expressed in HEK293 cells at ACRObiosystems.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 53.8 kDa. The protein migrates as 85-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant HIV-1 [HIV-1/Clade B/C (CN54)] GP120 (Cat. No. GP4-V15227) at 5 μg/mL (100 μL/well) can bind Human CD4, Fc Tag (Cat. No. CD4-H5259) with a linear range of 1-39 ng/mL (QC tested).
Catalog No. 50-101-7914