Learn More
ACROBiosystems Recombinant Protein;Human ICAM-1 / CD54 Protein, Fc Tag;HEK293;200UG;IC1,ICAM1,ICAM-1,BB2,BB-2,CD54,CD-54,P3.58
Supplier:  ACROBiosystems IC1H5250200UG
Encompass_Preferred
Recombinant Protein;Human ICAM-1 / CD54 Protein, Fc Tag;200UG;Human ICAM-1, Fc Tag (IC1-H5250) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Glu 480 (Accession # AAH15969).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.7 kDa. The protein migrates as 90-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ITGALB2, His tag at 5 μg/mL (100 μL/well) can bind Human ICAM-1, Fc Tag (Cat. No. IC1-H5250) with a linear range of 0.082-1.28 μg/mL (Routinely tested).
Catalog No. 50-101-7982