Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human LILRB2 / CD85d / ILT4 Protein;HEK293;100UG;LI2,LILRB2,ILT4,LIR2,MIR10,CD85d,LIR-2,ILT-4,MIR-10  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner LI2H5220100UG
Recombinant Protein;Human LILRB2 / CD85d / ILT4 Protein;100UG;Human LILRB2, His Tag (LI2-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # AAH36827).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-8238