Learn More
ACROBiosystems Recombinant Protein;Human Neuropilin-1 / NRP1 / CD304 Protein, Fc Tag;HEK293;50UG;NR1,NRP1,Neuropilin-1,NRP,VEGF165R,CD304,NRP-1
Supplier:  ACROBiosystems NR1H525250UG
Encompass_Preferred
Recombinant Protein;Human Neuropilin-1 / NRP1 / CD304 Protein, Fc Tag;50UG;Human NRP1, Fc Tag (NR1-H5252) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 96.5 kDa. The protein migrates as 105-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Human NRP1, Fc Tag (Cat. No. NR1-H5252) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-101-8343